siRNA Details
| Gene Symbol: Zfp446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168562.1 | NP_001162034.1 | 3 | 883 | ||
| NM_175558.4 | NP_780767.1 | 3 | 777 | |||
| GenBank mRNA | AK041761.1 | - | 3 | 779 | ||
| AK044666.1 | - | 2 | 1619 | |||
| AK138378.1 | - | 1 | 62 | |||
| AK142766.1 | - | 2 | 1272 | |||
| BC036728.1 | - | 2 | 729 | |||
| BC048909.1 | - | 3 | 719 | |||
| BC117850.1 | - | 3 | 890 | |||

Zfp446
Genomic Map