siRNA Details
| Gene Symbol: Tmem169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175564.4 | NP_780773.1 | 2 | 432 | ||
| GenBank mRNA | AK034935.1 | - | 2 | 437 | ||
| AK043681.1 | - | 2 | 433 | |||
| AK084021.1 | - | 2 | 411 | |||
| AK149157.1 | - | 1 | 742 | |||
| BC116730.1 | - | 1 | 330 | 40126049 | ||
| BC117087.1 | - | 1 | 330 | 40126406 | ||

Tmem169
Genomic Map