siRNA Details
| Gene Symbol: Plin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113471.1 | NP_001106942.1 | 9 | 1708 | ||
| NM_175640.2 | NP_783571.2 | 9 | 1764 | |||
| GenBank mRNA | AK031445.1 | - | 8 | 1689 | ||
| BC037748.1 | - | 2 | 482 | |||
| BC096685.1 | - | 9 | 1709 | 1448913 | ||

Plin1
Genomic Map