siRNA Details
| Gene Symbol: Kctd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175519.5 | NP_780728.3 | 2 | 2180 | ||
| GenBank mRNA | AK042569.1 | - | 2 | 2036 | ||
| AK043351.1 | - | 2 | 2150 | |||
| AK045439.1 | - | 2 | 2033 | |||
| AK047519.1 | - | 2 | 2182 | |||
| AY615967.1 | - | 2 | 2182 | |||

Kctd8
Genomic Map