siRNA Details
| Gene Symbol: Gphb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175644.3 | NP_783575.2 | 3 | 823 | ||
| GenBank mRNA | AK078727.1 | - | 3 | 827 | ||
| BC139462.1 | - | 3 | 763 | 8862484 | ||
| BC139463.1 | - | 3 | 763 | 8862485 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gphb5| Gene Symbol: Gphb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175644.3 | NP_783575.2 | 3 | 823 | ||
| GenBank mRNA | AK078727.1 | - | 3 | 827 | ||
| BC139462.1 | - | 3 | 763 | 8862484 | ||
| BC139463.1 | - | 3 | 763 | 8862485 | ||