siRNA Details
| Gene Symbol: Npsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175678.3 | NP_783609.1 | 10 | 3701 | ||
| GenBank mRNA | AK033957.1 | - | 10 | 3702 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npsr1| Gene Symbol: Npsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175678.3 | NP_783609.1 | 10 | 3701 | ||
| GenBank mRNA | AK033957.1 | - | 10 | 3702 | ||
Set Membership: |
|