siRNA Details
| Gene Symbol: Plxna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175750.3 | NP_786926.2 | 2 | 1126 | ||
| GenBank mRNA | AB073228.1 | - | 2 | 1125 | ||
| BC141298.1 | - | 2 | 424 | 9055910 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plxna4| Gene Symbol: Plxna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175750.3 | NP_786926.2 | 2 | 1126 | ||
| GenBank mRNA | AB073228.1 | - | 2 | 1125 | ||
| BC141298.1 | - | 2 | 424 | 9055910 | ||