siRNA Details
| Gene Symbol: Hps6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176785.3 | NP_789742.2 | 1 | 1211 | ||
| GenBank mRNA | AF536239.1 | - | 1 | 1156 | ||
| AK030582.1 | - | 1 | 1182 | |||
| AK040849.1 | - | 1 | 1158 | |||
| AK159588.1 | - | 1 | 1177 | |||

Hps6
Genomic Map