siRNA Details
| Gene Symbol: Exph5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176846.3 | NP_789816.2 | 1 | 140 | ||
| GenBank mRNA | AB098163.1 | - | 1 | 140 | ||
| BC120905.1 | - | 1 | 127 | 40087301 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Exph5| Gene Symbol: Exph5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176846.3 | NP_789816.2 | 1 | 140 | ||
| GenBank mRNA | AB098163.1 | - | 1 | 140 | ||
| BC120905.1 | - | 1 | 127 | 40087301 | ||