siRNA Details
| Gene Symbol: Ubald2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176902.3 | NP_795876.1 | 3 | 896 | ||
| GenBank mRNA | AK050732.1 | - | 3 | 777 | ||
| AK144901.1 | - | 3 | 901 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubald2| Gene Symbol: Ubald2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176902.3 | NP_795876.1 | 3 | 896 | ||
| GenBank mRNA | AK050732.1 | - | 3 | 777 | ||
| AK144901.1 | - | 3 | 901 | |||