siRNA Details
| Gene Symbol: Nwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176940.5 | NP_795914.3 | 5 | 997 | ||
| GenBank mRNA | AK034868.1 | - | 696 | |||
| AK038792.1 | - | 4 | 689 | |||
| AK087622.1 | - | 5 | 998 | |||
| AK162888.1 | - | 5 | 992 | |||
| BC082552.1 | - | 5 | 937 | |||
| BC144786.1 | - | 3 | 587 | 9053334 | ||
| BC144787.1 | - | 3 | 587 | |||
| BC145703.1 | - | 3 | 587 | 40131006 | ||

Nwd1
Genomic Map