siRNA Details
| Gene Symbol: 6430573F11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176952.4 | NP_795926.2 | 3 | 314 | ||
| GenBank mRNA | AK032504.1 | - | 5 | 1032 | ||
| AK038497.1 | - | 5 | 754 | |||
| AK039253.1 | - | 4 | 664 | |||
| AK141601.1 | - | 688 | ||||
| AK220392.1 | - | 1 | 167 | |||
| BC052184.1 | - | 3 | 314 | 6336362 | ||

6430573F11Rik
Genomic Map