siRNA Details
| Gene Symbol: Btn2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289614.1 | NP_001276543.1 | 3 | 593 | ||
| NM_001289615.1 | NP_001276544.1 | 3 | 534 | |||
| NM_175938.3 | NP_787952.2 | 3 | 593 | |||
| GenBank mRNA | AK051061.1 | - | 3 | 580 | ||
| AY177686.1 | - | 3 | 576 | |||
| BC139760.1 | - | 3 | 420 | 40058494 | ||
| BC141341.1 | - | 3 | 566 | 9055953 | ||

Btn2a2
Genomic Map