siRNA Details
| Gene Symbol: A730018C14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042713.1 | - | 3 | 1289 | ||
| BC172018.1 | - | 1 | 970 | 9054304 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A730018C14Rik| Gene Symbol: A730018C14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042713.1 | - | 3 | 1289 | ||
| BC172018.1 | - | 1 | 970 | 9054304 | ||