siRNA Details
| Gene Symbol: 9330132A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033982.1 | - | 1313 | |||
| AK083197.1 | - | 1090 | ||||
| BC098197.1 | - | 1041 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9330132A10Rik| Gene Symbol: 9330132A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033982.1 | - | 1313 | |||
| AK083197.1 | - | 1090 | ||||
| BC098197.1 | - | 1041 | ||||