siRNA Details
| Gene Symbol: Fsd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195284.1 | NP_001182213.1 | 2, 3 | 246 | ||
| NM_176966.4 | NP_795940.1 | 2, 3 | 246 | |||
| GenBank mRNA | AK038893.1 | - | 2, 3 | 224 | ||
| AK052914.1 | - | 2, 3 | 289 | |||
| AK140020.1 | - | 2, 3 | 230 | |||
| BB662298.1 | - | 2, 3 | 284 | |||
| BC130265.1 | - | 2, 3 | 162 | 40142990 | ||
| CJ115006.1 | - | 2, 3 | 247 | |||

Fsd1l
Genomic Map