siRNA Details
| Gene Symbol: Usp37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176972.4 | NP_795946.2 | 4 | 519 | ||
| GenBank mRNA | AK030013.1 | - | 4 | 520 | ||
| AK035640.1 | - | 3 | 503 | |||
| BC114208.1 | - | 3 | 483 | |||
| BC139091.1 | - | 4 | 461 | 8862113 | ||

Usp37
Genomic Map