siRNA Details
| Gene Symbol: Fsd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176966.4 | NP_795940.1 | 4 | 1505 | ||
| GenBank mRNA | AK038893.1 | - | 4 | 1484 | ||
| AK140020.1 | - | 4 | 1489 | |||
| BC130265.1 | - | 4 | 1421 | 40142990 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fsd1l| Gene Symbol: Fsd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176966.4 | NP_795940.1 | 4 | 1505 | ||
| GenBank mRNA | AK038893.1 | - | 4 | 1484 | ||
| AK140020.1 | - | 4 | 1489 | |||
| BC130265.1 | - | 4 | 1421 | 40142990 | ||