siRNA Details
| Gene Symbol: A430010J10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039826.1 | - | 1474 | |||
| AK083337.1 | - | 1 | 2932 | |||
| BB208751.1 | - | 1 | 78 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A430010J10Rik| Gene Symbol: A430010J10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039826.1 | - | 1474 | |||
| AK083337.1 | - | 1 | 2932 | |||
| BB208751.1 | - | 1 | 78 | |||