siRNA Details
Gene Symbol: Topbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176979.5 | NP_795953.2 | 5 | 505 | ||
GenBank mRNA | AK084654.1 | - | 5 | 608 | ||
AK085031.1 | - | 5 | 644 | |||
AK129104.1 | - | 5 | 617 | |||
BC049797.1 | - | 5 | 505 | 6510112 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
