siRNA Details
| Gene Symbol: Fbxo48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176982.2 | NP_795956.1 | 3, 4 | 1507 | ||
| GenBank mRNA | AK037835.1 | - | 3, 4 | 1507 | ||
| AK041978.1 | - | 3, 4 | 814 | |||
| BC125589.1 | - | 2, 3 | 384 | 40130104 | ||
| BC125591.1 | - | 2, 3 | 384 | 40130106 | ||
| BC145147.1 | - | 1, 2 | 479 | |||

Fbxo48
Genomic Map