siRNA Details
| Gene Symbol: Simc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176987.4 | NP_795961.3 | 2 | 2789 | ||
| GenBank mRNA | AK028926.1 | - | 2 | 439 | ||
| AK052561.1 | - | 1 | 583 | |||
| AK166176.1 | - | 1 | 393 | |||
| BC059035.1 | - | 1 | 1048 | |||
| BC094550.1 | - | 1 | 600 | |||

Simc1
Genomic Map