siRNA Details
| Gene Symbol: Cobll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027225.1 | NP_081501.1 | 1, 2 | 271 | ||
| NM_177025.5 | NP_795999.2 | 1, 2 | 271 | |||
| GenBank mRNA | AK052533.1 | - | 1, 2 | 220 | ||
| AK052939.1 | - | 2, 3 | 202 | |||
| AK164624.1 | - | 2, 3 | 201 | |||
| AY308746.1 | - | 1, 2 | 271 | |||

Cobll1
Genomic Map