siRNA Details
| Gene Symbol: BC106179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041922.1 | - | 2 | 156 | ||
| AK161755.1 | - | 2 | 441 | |||
| BC106179.1 | - | 2 | 437 | 6509459 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC106179| Gene Symbol: BC106179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041922.1 | - | 2 | 156 | ||
| AK161755.1 | - | 2 | 441 | |||
| BC106179.1 | - | 2 | 437 | 6509459 | ||