siRNA Details
| Gene Symbol: Adrbk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285806.1 | NP_001272735.1 | 3 | 313 | ||
| NM_177078.4 | NP_796052.2 | 3 | 313 | |||
| GenBank mRNA | AK048763.1 | - | 3 | 278 | ||
| AK076533.1 | - | 3 | 211 | |||
| AK134672.1 | - | 3 | 290 | |||
| AK141186.1 | - | 3 | 291 | |||
| AK172513.1 | - | 3 | 265 | |||
| CJ104587.1 | - | 3 | 314 | |||

Adrbk2
Genomic Map