siRNA Details
| Gene Symbol: Neurl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081656.2 | NP_001075125.1 | 5 | 5080 | ||
| GenBank mRNA | AK030658.1 | - | 1 | 379 | ||
| AK053917.1 | - | 1 | 1711 | |||
| AK082655.1 | - | 1 | 1762 | |||
| BC150855.1 | - | 1 | 1501 | |||
| BC150866.1 | - | 1 | 1503 | |||

Neurl1b
Genomic Map