siRNA Details
Gene Symbol: Nfxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK051057.1 | - | 1 | 1370 | ||
AK085068.1 | - | 1 | 1365 | |||
AK164247.1 | - | 1 | 1984 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nfxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK051057.1 | - | 1 | 1370 | ||
AK085068.1 | - | 1 | 1365 | |||
AK164247.1 | - | 1 | 1984 |