siRNA Details
| Gene Symbol: Tmem229a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177013.3 | NP_795987.2 | 1 | 1516 | ||
| GenBank mRNA | AK087465.1 | - | 1 | 1486 | ||
| AK140056.1 | - | 1 | 1467 | |||
| AK161777.1 | - | 1 | 1690 | |||
| BC147241.1 | - | 1 | 1331 | 9056292 | ||
| BC147242.1 | - | 1 | 1331 | 9056287 | ||

Tmem229a
Genomic Map