siRNA Details
| Gene Symbol: EFNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001406.3 | NP_001397.1 | 2 | 540 | ||
| GenBank mRNA | AK314841.1 | - | 2 | 646 | ||
| BC042944.2 | - | 2 | 540 | 5286243 | ||
| U57001.1 | - | 2 | 540 | |||
| U62775.1 | - | 2 | 344 | |||
| U66406.1 | - | 2 | 756 | |||

EFNB3
Genomic Map