siRNA Details
| Gene Symbol: Vwc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177033.3 | NP_796007.1 | 2 | 1238 | ||
| GenBank mRNA | AB292670.1 | - | 1 | 641 | ||
| AK033944.1 | - | 2 | 1032 | |||
| AK044771.1 | - | 2 | 773 | |||
| AK141514.1 | - | 2 | 1239 | |||
| BC120564.1 | - | 1 | 700 | 40129487 | ||
| BC137825.1 | - | 1 | 701 | 8860844 | ||
| DQ421811.1 | - | 1 | 704 | |||

Vwc2
Genomic Map