siRNA Details
| Gene Symbol: Ceacam19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177036.5 | NP_796010.1 | 2 | 348 | ||
| GenBank mRNA | AK047942.1 | - | 2 | 335 | ||
| AK079290.1 | - | 2 | 349 | |||
| AK163794.1 | - | 2 | 336 | |||
| BC107227.1 | - | 2 | 276 | 40051960 | ||
| BC107228.2 | - | 2 | 276 | 40051962 | ||
| BC109333.3 | - | 2 | 276 | 40051723 | ||

Ceacam19
Genomic Map