siRNA Details
| Gene Symbol: Adrbk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285806.1 | NP_001272735.1 | 5 | 526 | ||
| NM_177078.4 | NP_796052.2 | 5 | 487 | |||
| GenBank mRNA | AK048763.1 | - | 5 | 452 | ||
| AK076533.1 | - | 5 | 385 | |||
| AK134672.1 | - | 5 | 464 | |||
| AK141186.1 | - | 5 | 505 | |||
| AK172513.1 | - | 5 | 439 | |||

Adrbk2
Genomic Map