siRNA Details
| Gene Symbol: Fam179a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177087.4 | NP_796061.2 | 18 | 2748 | ||
| GenBank mRNA | AK052197.1 | - | 3 | 2438 | ||
| AK076284.1 | - | 17 | 2460 | |||
| AK158647.1 | - | 18 | 2533 | |||
| BC132489.1 | - | 2 | 1407 | |||

Fam179a
Genomic Map