siRNA Details
| Gene Symbol: Slc22a30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177002.3 | NP_795976.1 | 6 | 1195 | ||
| GenBank mRNA | AF408394.1 | - | 1097 | |||
| AK040781.1 | - | 6 | 1198 | |||
| AK050438.1 | - | 6 | 1197 | |||
| AK052592.1 | - | 7 | 1293 | |||
| AK080180.1 | - | 6 | 1197 | |||
| BC016496.1 | - | 6 | 1149 | |||
| BC050265.1 | - | 6 | 1167 | 5044850 | ||
| BC132146.1 | - | 6 | 1133 | 40130423 | ||
| BC132336.1 | - | 6 | 1133 | 40130613 | ||

Slc22a30
Genomic Map