siRNA Details
| Gene Symbol: Ceacam19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177036.5 | NP_796010.1 | 7 | 2270 | ||
| GenBank mRNA | AK079290.1 | - | 7 | 2272 | ||
| AK163794.1 | - | 7 | 2258 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ceacam19| Gene Symbol: Ceacam19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177036.5 | NP_796010.1 | 7 | 2270 | ||
| GenBank mRNA | AK079290.1 | - | 7 | 2272 | ||
| AK163794.1 | - | 7 | 2258 | |||