siRNA Details
| Gene Symbol: Tmem91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290497.1 | NP_001277426.1 | 2, 3 | 375 | ||
| NM_001290498.1 | NP_001277427.1 | 2, 3 | 375 | |||
| NM_177102.4 | NP_796076.1 | 2, 3 | 375 | |||
| GenBank mRNA | AK043858.1 | - | 2, 3 | 328 | ||
| AK079312.1 | - | 2, 3 | 376 | |||
| BC058996.1 | - | 2, 3 | 324 | 5711063 | ||
| BE863748.1 | - | 1, 2 | 256 | |||
| BQ555196.1 | - | 2, 3 | 303 | |||

Tmem91
Genomic Map