siRNA Details
| Gene Symbol: Tmem251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177140.3 | NP_796114.1 | 2 | 538 | ||
| GenBank mRNA | AK032513.1 | - | 2 | 544 | ||
| AK052027.1 | - | 2 | 540 | |||
| AK133919.1 | - | 2 | 541 | |||
| AK144149.1 | - | 541 | ||||
| AK172131.1 | - | 2 | 163 | |||
| BC096606.1 | - | 2 | 526 | 5366962 | ||

Tmem251
Genomic Map