siRNA Details
| Gene Symbol: Csnk1g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152809.2 | NP_690022.2 | 2 | 824 | ||
| GenBank mRNA | AK049379.1 | - | 2 | 826 | ||
| AK080491.1 | - | 2 | 817 | |||
| CN454971.1 | - | 1 | 307 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csnk1g3| Gene Symbol: Csnk1g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152809.2 | NP_690022.2 | 2 | 824 | ||
| GenBank mRNA | AK049379.1 | - | 2 | 826 | ||
| AK080491.1 | - | 2 | 817 | |||
| CN454971.1 | - | 1 | 307 | |||