siRNA Details
| Gene Symbol: Ubn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177185.4 | NP_796159.3 | 3, 4 | 874 | ||
| GenBank mRNA | AK031359.1 | - | 4, 5 | 842 | ||
| AK051609.1 | - | 3, 4 | 241 | |||
| BC043095.1 | - | 122 | ||||
| BC051458.1 | - | 3, 4 | 849 | 6468017 | ||

Ubn2
Genomic Map