siRNA Details
| Gene Symbol: Ccdc171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081012.1 | NP_001074481.1 | 17 | 2841 | ||
| GenBank mRNA | AK039282.1 | - | 4 | 959 | ||
| BC172165.1 | - | 16 | 2632 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc171| Gene Symbol: Ccdc171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081012.1 | NP_001074481.1 | 17 | 2841 | ||
| GenBank mRNA | AK039282.1 | - | 4 | 959 | ||
| BC172165.1 | - | 16 | 2632 | |||