siRNA Details
| Gene Symbol: Iqcb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177128.4 | NP_796102.2 | 15 | 1983 | ||
| GenBank mRNA | AK078554.1 | - | 15 | 1976 | ||
| AK165220.1 | - | 15 | 1967 | |||
| BE652573.1 | - | 1 | 181 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqcb1| Gene Symbol: Iqcb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177128.4 | NP_796102.2 | 15 | 1983 | ||
| GenBank mRNA | AK078554.1 | - | 15 | 1976 | ||
| AK165220.1 | - | 15 | 1967 | |||
| BE652573.1 | - | 1 | 181 | |||