siRNA Details
| Gene Symbol: Pde4dip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031066.1 | - | 1 | 360 | ||
| AK051209.1 | - | 4 | 670 | |||
| AK083867.1 | - | 4 | 667 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pde4dip| Gene Symbol: Pde4dip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031066.1 | - | 1 | 360 | ||
| AK051209.1 | - | 4 | 670 | |||
| AK083867.1 | - | 4 | 667 | |||