siRNA Details
| Gene Symbol: Tmprss11g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177162.4 | NP_796136.2 | 5 | 443 | ||
| GenBank mRNA | AK036981.1 | - | 5 | 444 | ||
| BC118611.1 | - | 5 | 421 | 40107184 | ||
| BC118660.1 | - | 5 | 420 | 40107187 | ||
| CK627573.1 | - | 5 | 501 | |||

Tmprss11g
Genomic Map