siRNA Details
| Gene Symbol: 9930022D16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK027987.1 | - | 1, 2 | 218 | ||
| AK036894.1 | - | 1, 2 | 219 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_177179.1 | - | 219 | Chr. - - [-] on Build | ||

9930022D16Rik
Genomic Map