siRNA Details
| Gene Symbol: Tmem91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290497.1 | NP_001277426.1 | 4 | 699 | ||
| NM_001290498.1 | NP_001277427.1 | 4 | 677 | |||
| NM_177102.4 | NP_796076.1 | 4 | 847 | |||
| GenBank mRNA | AK079312.1 | - | 4 | 848 | ||
| BC058996.1 | - | 4 | 796 | 5711063 | ||

Tmem91
Genomic Map