siRNA Details
| Gene Symbol: Ccdc66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177111.3 | NP_796085.3 | 14 | 2005 | ||
| GenBank mRNA | AK087583.1 | - | 6 | 726 | ||
| AK158666.1 | - | 12 | 1523 | |||
| AK170006.1 | - | 12 | 1472 | |||
| BC070471.1 | - | 14 | 2005 | 30626499 | ||
| BC072583.1 | - | 6 | 1122 | 6403859 | ||

Ccdc66
Genomic Map