siRNA Details
| Gene Symbol: Csnk1g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049379.1 | - | 2 | 1062 | ||
| AK080491.1 | - | 2 | 1055 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csnk1g3| Gene Symbol: Csnk1g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049379.1 | - | 2 | 1062 | ||
| AK080491.1 | - | 2 | 1055 | |||