siRNA Details
| Gene Symbol: Tmem215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166009.1 | NP_001159481.1 | 2 | 1808 | ||
| NM_177175.4 | NP_796149.2 | 2 | 2750 | |||
| GenBank mRNA | AK044217.1 | - | 2 | 2752 | ||
| AK044627.1 | - | 2 | 2752 | |||

Tmem215
Genomic Map