siRNA Details
| Gene Symbol: Ubn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177185.4 | NP_796159.3 | 17 | 4330 | ||
| GenBank mRNA | AK051609.1 | - | 17 | 3703 | ||
| AK173339.1 | - | 5 | 5366 | |||
| BC051458.1 | - | 17 | 4302 | 6468017 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubn2| Gene Symbol: Ubn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177185.4 | NP_796159.3 | 17 | 4330 | ||
| GenBank mRNA | AK051609.1 | - | 17 | 3703 | ||
| AK173339.1 | - | 5 | 5366 | |||
| BC051458.1 | - | 17 | 4302 | 6468017 | ||