siRNA Details
| Gene Symbol: Fam19a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177233.5 | NP_796207.2 | 5 | 752 | ||
| GenBank mRNA | AK081531.1 | - | 5 | 735 | ||
| AK149355.1 | - | 5 | 755 | |||
| AY325123.1 | - | 5 | 535 | |||
| BC089490.1 | - | 5 | 712 | 30504398 | ||

Fam19a4
Genomic Map